Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin heat1KG
Catalog No. 502907968
Encompass
Bovine serum albumin is a protein which has a key role in regulating blood colloidal osmotic pressure. A water-soluble protein composed of 583 amino acids BSA has a calculated molecular weight of 66430 Daltons. Six -helices form three homologous domains of BSA. Depending on pH BSA undergoes reversible conformational isomerization. The native structure of the protein becomes reactive and flexible on heating.This BSA product is specifically evaluated and tested for the following properties.nbspVery low protease content / protease-free.nbspVery low fatty acid content / fatty acid-freeProtease-free BSA is especially useful for applications like.nbspEnzyme assays.nbspProtein-based assays.nbspProtease-sensitive techniques that includea. Enzyme immunoassay (EIA)b. Nucleic acid hybridizationc. Radioimmunoassay (RIA)
Catalog No. 50-290-7968 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A70301KG
Add to Cart
Add to Cart
Bovine serum albumin is a protein which has a key role in regulating blood colloidal osmotic pressure. A water-soluble protein composed of 583 amino acids BSA has a calculated molecular weight of 66430 Daltons. Six -helices form three homologous domains of BSA. Depending on pH BSA undergoes reversible conformational isomerization. The native structure of the protein becomes reactive and flexible on heating.This BSA product is specifically evaluated and tested for the following properties.nbspVery low protease content / protease-free.nbspVery low fatty acid content / fatty acid-freeProtease-free BSA is especially useful for applications like.nbspEnzyme assays.nbspProtein-based assays.nbspProtease-sensitive techniques that includea. Enzyme immunoassay (EIA)b. Nucleic acid hybridizationc. Radioimmunoassay (RIA)