Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin heat1KG
Catalog No. 502903985
Encompass
Bovine serum albumin (BSA) is a 66.4 kDa water-soluble protein composed of 583 amino acids. It has a single polypeptide chain with three homologous domains formed by six helices. Depending on pH it undergoes reversible conformational isomerization. The native structure of the protein becomes reactive and flexible on heating. BSA has been used as a model protein in various applications including immunodiagnostic procedures cell culture media and clinical chemistry. Serum albumins facilitates drug disposition and efficacy by maintaining osmotic blood pressure.
Catalog No. 50-290-3985 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A39121KG
Bovine serum albumin (BSA) is a 66.4 kDa water-soluble protein composed of 583 amino acids. It has a single polypeptide chain with three homologous domains formed by six helices. Depending on pH it undergoes reversible conformational isomerization. The native structure of the protein becomes reactive and flexible on heating. BSA has been used as a model protein in various applications including immunodiagnostic procedures cell culture media and clinical chemistry. Serum albumins facilitates drug disposition and efficacy by maintaining osmotic blood pressure.