Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin heat500G
Catalog No. 502907951
Encompass
Bovine serum albumin (BSA) is a serum protein with a calculated molecular weight of 66 430 Daltons BSA consists of three domains with two subdomains under each and is a helical globular and non-glycosylated protein with 17-disulfide bonds.BSA acts as a carrier protein for biomolecules like as fatty acids amino acids and steroids.
Catalog No. 50-290-7951 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A8022500G
Bovine serum albumin (BSA) is a serum protein with a calculated molecular weight of 66 430 Daltons BSA consists of three domains with two subdomains under each and is a helical globular and non-glycosylated protein with 17-disulfide bonds.BSA acts as a carrier protein for biomolecules like as fatty acids amino acids and steroids.