Learn More
Medchemexpress LLC Bovine Serum Albumin (low endotoxin, fatty acid free) | 9048-46-8 | 99.9% | 1 G
SDP
Catalog No. 5000409900
Encompass_Preferred
Bovine Serum Albumin (BSA) is a 583 amino acid globular protein and oligonucleotide binding protein composed of three homologous full α domains. It is designed for research applications, with specific activities observed in vitro and in vivo.
  • Blocks overall oligonucleotide association with cells.
  • Regulates the development of hamster embryos.
  • Can induce arthritis in rabbits.
  • Composed of three homologous full alpha domains.
  • Low endotoxin and fatty acid-free formulation.
  • High purity of 99.88%.
Catalog No. 50-004-09900 Supplier Medchemexpress LLC Supplier No. HYD0842A1G
Add to Cart
Add to Cart
Bovine Serum Albumin (BSA) is a 583 amino acid globular protein and oligonucleotide binding protein composed of three homologous full α domains. It is designed for research applications, with specific activities observed in vitro and in vivo.
  • Blocks overall oligonucleotide association with cells.
  • Regulates the development of hamster embryos.
  • Can induce arthritis in rabbits.
  • Composed of three homologous full alpha domains.
  • Low endotoxin and fatty acid-free formulation.
  • High purity of 99.88%.