Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin lyoph100G
Catalog No. 502907424
Encompass
Bovine serum albumin (BSA) is a non-glycosylated protein belonging to the serum albumins family. It is a globular protein with helix structure and 17-disulfide bonds. BSA has a molecular weight of 66 kDa consisting three domains and two subdomains under each. BSA shows structural similarity with human serum albumin.
Catalog No. 50-290-7424 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A2058100G
Bovine serum albumin (BSA) is a non-glycosylated protein belonging to the serum albumins family. It is a globular protein with helix structure and 17-disulfide bonds. BSA has a molecular weight of 66 kDa consisting three domains and two subdomains under each. BSA shows structural similarity with human serum albumin.