Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin lyoph100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A4378100G
Encompass
Albumin is the most abundant plasma protein in humans. High levels of albumin are associated with dehydration. Albumin turnover is seen in infants with iron deficiency anemia. Serum albumin is a reliable prognostic indicator for liver disease. Oxidative damage of albumin is associated with advanced liver disease.
Catalog No. 50-290-7554