Learn More
Abcam Bovine Serum Albumin (nitrated ), 100UG
Catalog No. 502707031
Encompass_Preferred
Bovine Serum Albumin (nitrated ) is a Cow Full Length protein, in the 1 to 607 aa range, expressed in Native, with >96% purity and suitable for SDS-PAGE, WB.
Protein/ Peptide/ Enzyme Synonyms: Albumin, BSA, ALB
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7031 Supplier Abcam Supplier No. AB131379100UG
Bovine Serum Albumin (nitrated ) is a Cow Full Length protein, in the 1 to 607 aa range, expressed in Native, with >96% purity and suitable for SDS-PAGE, WB.
Protein/ Peptide/ Enzyme Synonyms: Albumin, BSA, ALB
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement