Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin solut
Catalog No. 502899243
Encompass
Bovine serum albumin (BSA)nbspis a member of the serum albuminnbspfamily. It is anbspglobular helical and non-glycosylatednbspprotein. BSA is a single-chain protein and consists ofnbspthree domains with two sub-domains under eachItnbsphas 17-disulfidenbspbonds.
Catalog No. 50-289-9243 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A329950ML
Bovine serum albumin (BSA)nbspis a member of the serum albuminnbspfamily. It is anbspglobular helical and non-glycosylatednbspprotein. BSA is a single-chain protein and consists ofnbspthree domains with two sub-domains under eachItnbsphas 17-disulfidenbspbonds.