Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin solut
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A797912X50ML
Encompass
BSA is a single polypeptide chain consisting of about 583 amino acid residues and no carbohydrates. At pH 5-7 it contains 17 intrachain disulfide bridges and 1 sulfhydryl group.
Catalog No. 50-290-6988