Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin solut1L
Catalog No. 502907575
Encompass
Bovine Serum Albumin (BSA) has three homologous domains each subdivided it two domain (A and B). It is a helical globular protein and shares structural features with human serum albumin.
Catalog No. 50-290-7575 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A92051L
Bovine Serum Albumin (BSA) has three homologous domains each subdivided it two domain (A and B). It is a helical globular protein and shares structural features with human serum albumin.