Learn More
Biomatik Corporation Bovine Sorbitol Dehydrogenase (SDH) ELISA Kit
Supplier:  Biomatik Corporation EKU07414
Encompass_Preferred
Bovine Sorbitol Dehydrogenase (SDH) ELISA Kit for detecting SDH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.13ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q58D31. Gene ID: 508954. Target Synonyms: SORD; L-iditol 2-dehydrogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7126