Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine srum Albumin solut250ML
Catalog No. 502902702
Encompass
Bovine serum albumin (BSA) belongs to the serum albumin family. It comprises three domains with two sub-domains under each. It is an helical globular and non-glycosylated protein. BSA has 17-disulfide bonds.
Catalog No. 50-290-2702 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A3174250ML
Bovine serum albumin (BSA) belongs to the serum albumin family. It comprises three domains with two sub-domains under each. It is an helical globular and non-glycosylated protein. BSA has 17-disulfide bonds.