Learn More
Biomatik Corporation Bovine Sulfite Oxidase (SUOX) ELISA Kit
Supplier:  Biomatik Corporation EKU07496
Encompass_Preferred
Bovine Sulfite Oxidase (SUOX) ELISA Kit for detecting SUOX in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F6RUR3. Gene ID: 509837. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7576