Learn More
Cusabio Technology LLC BOVINE TGF beta 1 ELISA KIT
Catalog No. 5000048924
Encompass_Preferred
This ELISA kit is designed for the quantitative detection of Bovine transforming growth factor B1 (TGF-B1) in serum plasma and cell culture supernatants Utilizing a sandwich assay format it offers a broad detection range of 6 25 ng/mL to 400 ng/mL with a sensitivity of 1 56 ng/mL enabling precise measurement of low-abundance targets The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours making it efficient for routine laboratory workflows Absorbance is measured at 450 nm to determine TGF-B1 concentrations ensuring reliable and reproducible results This kit is optimized for research applications requiring accurate quantification of TGFB1 in biological matrices
Catalog No. 50-000-48924 Supplier Cusabio Technology LLC Supplier No. CSBE14208B
This ELISA kit is designed for the quantitative detection of Bovine transforming growth factor B1 (TGF-B1) in serum plasma and cell culture supernatants Utilizing a sandwich assay format it offers a broad detection range of 6 25 ng/mL to 400 ng/mL with a sensitivity of 1 56 ng/mL enabling precise measurement of low-abundance targets The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours making it efficient for routine laboratory workflows Absorbance is measured at 450 nm to determine TGF-B1 concentrations ensuring reliable and reproducible results This kit is optimized for research applications requiring accurate quantification of TGFB1 in biological matrices