Learn More
Cusabio Technology LLC BOVINE TGF beta 2 ELISA KIT
Catalog No. 5000048756
Encompass_Preferred
This kit detects TGFB2 in bovine serum plasma and tissue homogenates with a sensitivity of 6 25 ng/mL Designed for quantitative analysis it employs a competitive ELISA format providing reliable measurements within the range of 6 25 ng/mL to 100 ng/mL The assay requires a sample volume of 50-100 L and can be completed in 1-5 hours Detection is performed at 450 nm ensuring accurate and reproducible results for researchers studying TGFB2 in various biological matrices
Catalog No. 50-000-48756 Supplier Cusabio Technology LLC Supplier No. CSBE14209B
This kit detects TGFB2 in bovine serum plasma and tissue homogenates with a sensitivity of 6 25 ng/mL Designed for quantitative analysis it employs a competitive ELISA format providing reliable measurements within the range of 6 25 ng/mL to 100 ng/mL The assay requires a sample volume of 50-100 L and can be completed in 1-5 hours Detection is performed at 450 nm ensuring accurate and reproducible results for researchers studying TGFB2 in various biological matrices