Learn More
Pacific Hemostasis™ Bovine Thrombin 200 (100 NIH Units/mL), 10 x 2mL
Screen and quantify plasma fibrinogen based on the Clauss method.
Description
Screen and quantify plasma fibrinogen based on the Clauss method with Thermo Scientific™ Pacific Hemostasis™ Fibrinogen/Fibrin Analysis.
Fibrinogen Assay Set:- Quantifies plasma fibrinogen levels
- Detects acquired and congenital hemorrhagic disorders
- Components available separately
- Provides qualitative determination
- Useful in screening for fibrinogen disorders by measuring the formation of the fibrin clot
- Reconstituted stability: 8 hours at 2° to 8°C
Ordering Information:
Products can be purchased as a set or individually.