Learn More
Pacific Hemostasis™ Bovine Thrombin 500 (100 NIH Units/mL), 10 x 5mL
Fisherbrand™ Pacific Hemostasis Fibrinogen/Fibrin Analysis is a complete line of assays used to screen and quantify plasma fibrinogen based on the Clauss method.
Catalog No. 23-306291
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Thermo Scientific™ Pacific Hemostasis Fibrinogen/Fibrin Analysis is a complete line of assays used to screen and quantify plasma fibrinogen based on the Clauss method.
  • Fibrinogen Assay Set - quantifies plasma fibrinogen levels
  • Detects acquired and congenital hemorrhagic disorders
  • Components available separately
  • Thrombin Time Assay
  • provides a qualitative determination
  • Useful in screening for fibrinogen disorders by measuring the formation of the fibrin clot
  • Reconstituted stability: 8 hours at 2° to 8°C
Ordering Information:
Products can be purchased as a set or individually.
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions