Learn More
American Research Products Inc BoVine Thrombin antithrombin I
Supplier:  American Research Products Inc CSBE14133B
Encompass
BoVine Thrombin-antithrombin III CompleX TAT-III ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma Method Competitive
Catalog No. 50-003-30105