Learn More
Biomatik Corporation Bovine Thymic Factor (TF) ELISA Kit
Supplier:  Biomatik Corporation EKU11581
Encompass_Preferred
Bovine Thymic Factor (TF) ELISA Kit for detecting TF in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 37.0-3,000pg/ml. Sensitivity: 13.3pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: FTS; Facteur Thymique Serique; Thymulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5782