Learn More
American Research Products Inc BoVine thyroXine T4 ELISA Kit
Catalog No. 5000329045
Encompass
BoVine thyroXine T4 ELISA Kit from CUSABIO Detection Range 20 ng/mL-320 ng/mL Sensitivity 20 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29045 Supplier American Research Products Inc Supplier No. CSBE13050B
BoVine thyroXine T4 ELISA Kit from CUSABIO Detection Range 20 ng/mL-320 ng/mL Sensitivity 20 ng/mL Sample serum plasma tissue homogenates Method Competitive