Learn More
Biomatik Corporation Bovine Thyroxine, T4 Elisa Kit
Supplier:  Biomatik Corporation EKC31021
Encompass_Preferred
Bovine Thyroxine, T4 Elisa Kit for detecting Thyroxine, T4 in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 20-320ng/ml. Sensitivity: 20ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5151