Learn More
Biomatik Corporation Bovine Tissue Inhibitors Of Metalloproteinase 3 (TIMP3) ELISA Kit
Supplier:  Biomatik Corporation EKU07720
Encompass_Preferred
Bovine Tissue Inhibitors Of Metalloproteinase 3 (TIMP3) ELISA Kit for detecting TIMP3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P79121. Gene ID: 282094. Target Synonyms: SFD; TIMP Metallopeptidase Inhibitor 3; Metallopeptidase Inhibitor 3; Sorsby Fundus Dystrophy; Pseudoinflammatory. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6876