Learn More
Biomatik Corporation Bovine Tissue Inhibitors Of Metalloproteinase 2 (TIMP2) ELISA Kit
Supplier:  Biomatik Corporation EKU07714
Encompass_Preferred
Bovine Tissue Inhibitors Of Metalloproteinase 2 (TIMP2) ELISA Kit for detecting TIMP2 in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 62.50-4000pg/ml. Sensitivity: 26.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16368. Gene ID: 282093. Target Synonyms: CSC-21K; TIMP Metallopeptidase Inhibitor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7233