Learn More
Biomatik Corporation Bovine Tissue Inhibitors Of Metalloproteinase 4 (TIMP4) ELISA Kit
Supplier:  Biomatik Corporation EKU07730
Encompass_Preferred
Bovine Tissue Inhibitors Of Metalloproteinase 4 (TIMP4) ELISA Kit for detecting TIMP4 in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O97563. Gene ID: 317694. Target Synonyms: TIMP Metallopeptidase Inhibitor 4; Tissue Inhibitor of Metalloproteinases 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8731