Learn More
Sino Biological Bovine TMPRSS15 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10002MM0120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Bovine Enterokinase / PRSS7 (Catalog#10002-B08Y; NP_776864.1; Ile801-His1035). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3287