Learn More
Sino Biological Anti-Enterokinase Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10002T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Bovine Enterokinase / PRSS7 ( Catalog#10002-B08Y; NP_776864.1; Ile801-His1035). Enterokinase / PRSS7 specific IgG was purified by Bovine Enterokinase / PRSS7 affinity chromatography.
Catalog No. 50-241-1593