Learn More
Kingfisher Biotech Inc Bovine TNF alpha Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0055B025
Encompass
The Bovine TNF alpha yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Bovine TNF alpha applications are for cell culture, ELISA standard, and Western Blot Control. The Bovine TNF alpha yeast-derived recombinant protein can be purchased in multiple sizes. Bovine TNF alpha Specifications: (Molecular Weight: 17.4 kDa) (Amino Acid Sequence: LRSSSQASSN KPVAHVVADI NSPGQLRWWD SYANALMANG VKLEDNQLVV PADGLYLIYS QVLFRGQGCP STPLFLTHTI SRIAVSYQTK VNILSAIKSP CHRETPEWAE AKPWYEPIYQ GGVFQLEKGD RLSAEINLPD YLDYAESGQV YFGIIAL) (Gene ID: 280943). For research use only. Made in the USA
Catalog No. 50-241-3555