Learn More
Raybiotech Inc Bovine TNF alpha ELISA Kit
Supplier:  Raybiotech Inc ELBTNFA1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.1 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects bovine TNF-alpha. Other species not determined
Catalog No. 50-194-8874