Learn More
American Research Products Inc BoVine transforming growth fac
Catalog No. 5000329451
Encompass
BoVine transforming growth factors b2 TGFb2 ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-100 ng/mL Sensitivity 6 25 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29451 Supplier American Research Products Inc Supplier No. CSBE14209B
BoVine transforming growth factors b2 TGFb2 ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-100 ng/mL Sensitivity 6 25 ng/mL Sample serum plasma tissue homogenates Method Competitive