Learn More
American Research Products Inc BoVine Transthyretin TTR ELI
Catalog No. 5000327588
Encompass
BoVine Transthyretin (TTR) ELISA kit from CUSABIO Detection Range 312 5 ng/mL-5000 ng/mL Sensitivity 312 5 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27588 Supplier American Research Products Inc Supplier No. CSBEL025270BO
BoVine Transthyretin (TTR) ELISA kit from CUSABIO Detection Range 312 5 ng/mL-5000 ng/mL Sensitivity 312 5 ng/mL Sample serum plasma tissue homogenates Method Competitive