Learn More
American Research Products Inc BoVine Tri iodothyronine T3 E
Catalog No. 5000329296
Encompass
BoVine Tri-iodothyronine T3 ELISA Kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 5 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29296 Supplier American Research Products Inc Supplier No. CSBE13049B
BoVine Tri-iodothyronine T3 ELISA Kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 5 ng/mL Sample serum plasma tissue homogenates Method Competitive