Learn More
Biomatik Corporation Bovine Tri-Iodothyronine, T3 Elisa Kit
Supplier:  Biomatik Corporation EKC31022
Encompass_Preferred
Bovine Tri-Iodothyronine, T3 Elisa Kit for detecting Tri-Iodothyronine, T3 in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.5-8ng/ml. Sensitivity: 0.5ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4980