Learn More
Worthington Biochemical Corporation BOVINE TRYPSIN DIALYZED AGAIN
SDP
Catalog No. NC9117581
Encompass
Trypsin, 3X Crystallized, Dialyzed Against 1mM HCl and Lyophilized, Store at 2 deg. to 8 deg.C, Source: Bovine Pancreas, Minimum Activity: 180 units/mg Protein, 100mg
Catalog No. NC9117581 Supplier Worthington Biochemical Corporation Supplier No. LS003708
Trypsin, 3X Crystallized, Dialyzed Against 1mM HCl and Lyophilized, Store at 2 deg. to 8 deg.C, Source: Bovine Pancreas, Minimum Activity: 180 units/mg Protein, 100mg