Learn More
American Research Products Inc BoVine Ubiquitin like protein
Catalog No. 5000326938
Encompass
BoVine Ubiquitin-like protein ISG15 (ISG15) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26938 Supplier American Research Products Inc Supplier No. CSBEL011843BO
BoVine Ubiquitin-like protein ISG15 (ISG15) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich