Learn More
American Research Products Inc BoVine Vascular endothelial gr
Supplier:  American Research Products Inc CSBEL025833BO
Encompass
BoVine Vascular endothelial growth factor A (VEGFA) ELISA kit from CUSABIO Detection Range 47 pg/mL-3000 pg/mL Sensitivity 11 75 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27592