Learn More
Vmrd Inc Bovine Herpesvirus Type 1 (BHV-1/IBR) FITC Conjugate 10 ml
SDP
Catalog No. NC1421888
Encompass
Anti-Bovine Herpesvirus Type 1/Infectious Bovine Rhinotracheitis (BHV/1/IBR) polyclonal antiserum conjugated to fluorescein isothiocyanate. Caprine origin. Useful for the detection of viruses in animal tissues or cell cultures. Does not react with other common bovine viruses.
Catalog No. NC1421888 Supplier Vmrd Inc Supplier No. CJFIBR50X
Anti-Bovine Herpesvirus Type 1/Infectious Bovine Rhinotracheitis (BHV/1/IBR) polyclonal antiserum conjugated to fluorescein isothiocyanate. Caprine origin. Useful for the detection of viruses in animal tissues or cell cultures. Does not react with other common bovine viruses.