Learn More
BURKLE INC Bowl stainless steel, deep form, 15.75x7.87x3.94 in. (400x200x100mm)
Catalog No. 5000025920
Encompass_Preferred
The stainless steel bowl is suitable for a wide variety of applications, e.g. for storing, carrying or cleaning laboratory cutlery or samplers. The smooth surface of the tray makes it easy to clean.
The instrument tray has a rectangular shape with a high edge and is very stable. Ideal for use in the pharmaceutical and chemical sectors as well as the food and cosmetics sectors.
Catalog No. 50-000-25920 Supplier BURKLE INC Supplier No. 42080040
The stainless steel bowl is suitable for a wide variety of applications, e.g. for storing, carrying or cleaning laboratory cutlery or samplers. The smooth surface of the tray makes it easy to clean.
The instrument tray has a rectangular shape with a high edge and is very stable. Ideal for use in the pharmaceutical and chemical sectors as well as the food and cosmetics sectors.