Learn More
BURKLE INC Bowl stainless steel, shallow form, 11.81x7.87x1.97 in. (300x200x50mm)
Catalog No. 5000025982
Encompass_Preferred
The robust stainless steel bowl is ideal for cleaning, disinfecting or sterilizing instruments as well as for placing and carrying different devices or samples. Due to the smooth surface, the instrument tray is easy to clean.
The rectangular bowl has a low edge and can be used in the various areas of pharmacy, chemistry, cosmetics and food.
Catalog No. 50-000-25982 Supplier BURKLE INC Supplier No. 42080022
The robust stainless steel bowl is ideal for cleaning, disinfecting or sterilizing instruments as well as for placing and carrying different devices or samples. Due to the smooth surface, the instrument tray is easy to clean.
The rectangular bowl has a low edge and can be used in the various areas of pharmacy, chemistry, cosmetics and food.