Learn More
Medchemexpress LLC Bowman-birk inhibitor | 37330-34-0 | 5 MG
SDP
Catalog No. 5000396601
Encompass_Preferred
The Bowman-Birk inhibitor is a highly cross-linked protein with seven disulfide bridges. It has spatially distinct domains designed to inhibit both trypsin and chymotrypsin, making it a significant plant protease inhibitor.
  • Highly cross-linked protein with seven disulfide bridges
  • Distinct domains to inhibit both trypsin and chymotrypsin
  • Functions as a plant protease inhibitor
  • Exhibits anticarcinogenic properties
Catalog No. 50-003-96601 Supplier Medchemexpress LLC Supplier No. HYP30265MG
The Bowman-Birk inhibitor is a highly cross-linked protein with seven disulfide bridges. It has spatially distinct domains designed to inhibit both trypsin and chymotrypsin, making it a significant plant protease inhibitor.
  • Highly cross-linked protein with seven disulfide bridges
  • Distinct domains to inhibit both trypsin and chymotrypsin
  • Functions as a plant protease inhibitor
  • Exhibits anticarcinogenic properties