Learn More
Medchemexpress LLC Bowman-birk inhibitor | 37330-34-0 | 50 MG
SDP
Catalog No. 5000398433
Encompass_Preferred
The Bowman-Birk inhibitor is a highly cross-linked protein featuring seven disulfide bridges. It possesses spatially distinct domains specifically designed for the inhibition of both trypsin and chymotrypsin. This highlights its significant role as a plant protease inhibitor with anticarcinogenic properties.
  • Highly cross-linked protein structure
  • Contains seven disulfide bridges
  • Spatially distinct domains for inhibition
  • Specifically inhibits trypsin
  • Specifically inhibits chymotrypsin
  • Functions as a plant protease inhibitor
  • Possesses anticarcinogenic properties
  • Relevant for various research applications
Catalog No. 50-003-98433 Supplier Medchemexpress LLC Supplier No. HYP302650MG
The Bowman-Birk inhibitor is a highly cross-linked protein featuring seven disulfide bridges. It possesses spatially distinct domains specifically designed for the inhibition of both trypsin and chymotrypsin. This highlights its significant role as a plant protease inhibitor with anticarcinogenic properties.
  • Highly cross-linked protein structure
  • Contains seven disulfide bridges
  • Spatially distinct domains for inhibition
  • Specifically inhibits trypsin
  • Specifically inhibits chymotrypsin
  • Functions as a plant protease inhibitor
  • Possesses anticarcinogenic properties
  • Relevant for various research applications