Learn More
Alkali Scientific BowTie™ Biohazard Bags, Autoclave Bag with Drawstring, PE, 25 x 35", with Marking Area and Sterilization Indicator, 100/pk
SDP
Catalog No. 502335803
Encompass_Preferred
The BowTie™ Biohazard Bags exclusive drawstring loops provide an instant cinch closure and easy removal from bag stands. These drawstring loops allow for venting during sterilization and offer an easy and safe way to remove bags from a hot autoclave. Bags are clearly marked with a biohazard symbol. The BowTie Biohazard Bag is made of 2.25 mil HDPE and is suitable for autoclaving in 121°C cycles. They have a color changing sterilixation indicator that turns from white to black after sterilization. Dimenstion: 25 x 35". 12-14 gallon capacity.
Catalog No. 50-233-5803 Supplier Alkali Scientific Supplier No. A3003R
The BowTie™ Biohazard Bags exclusive drawstring loops provide an instant cinch closure and easy removal from bag stands. These drawstring loops allow for venting during sterilization and offer an easy and safe way to remove bags from a hot autoclave. Bags are clearly marked with a biohazard symbol. The BowTie Biohazard Bag is made of 2.25 mil HDPE and is suitable for autoclaving in 121°C cycles. They have a color changing sterilixation indicator that turns from white to black after sterilization. Dimenstion: 25 x 35". 12-14 gallon capacity.