Learn More
TWD Scientific LLC BioX 100-Well Hinged Cardboard Freezer Box, 1 Inch, Assorted Colors. Case of 20
SDP
Catalog No. 502274271 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX 1 inch 100 well water resistant id-color™ cardboard freezer boxes with hinged lid and divider made of PP material. Boxes have marked alphanumeric grid for identification purposes. Boxes come in assorted colors (red, blue, green, yellow and white). 5 boxes/pack, 4 packs/case, price/case
This item is not returnable. View return policy
Catalog No. 50-227-4271 Supplier TWD Scientific LLC Supplier No. BX908100
BioX 1 inch 100 well water resistant id-color™ cardboard freezer boxes with hinged lid and divider made of PP material. Boxes have marked alphanumeric grid for identification purposes. Boxes come in assorted colors (red, blue, green, yellow and white). 5 boxes/pack, 4 packs/case, price/case