Learn More
Thomas Scientific CryostoreT Storage Box, 3 to 5 mL, 81-Place, Red
SDP
Catalog No. 501461036
Encompass_Preferred
Made of polycarbonate Color your world with a wide variety of Cryostore™ Storage Boxes for sizes from 1.2 mL to 5 mL. Made of extra strong polycarbonate, these durable cryogenic storage boxes are designed to be used at temperatures between -196°C and +121°C and are autoclavable at 120°C, 15 psig (1 bar) for 20 minutes. Different models are available to accommodate either 25, 81 or 100 Cryovial tubes from 1.2 mL to 5 mL. A transparent cover allows the user to see the contents of the box, and is keyed to the base to prevent misalignment. Printed with a series of squares (numbered from 1 to 25, 1 to 42, 1 to 81, or 1 to 100), surface accepts writing with markers, facilitating inventory control. A unique color coding system uses colored plastic grids to separate the cover from the base on the 25, 42 and 81-place boxes. Those made to accept 100 tubes (series 2100) have a colored base instead of a grid. Removal of vials facilitated by an innovative vial picker supplied
Catalog No. 50-146-1036 Supplier Thomas Scientific Supplier No. 1167U01
Made of polycarbonate Color your world with a wide variety of Cryostore™ Storage Boxes for sizes from 1.2 mL to 5 mL. Made of extra strong polycarbonate, these durable cryogenic storage boxes are designed to be used at temperatures between -196°C and +121°C and are autoclavable at 120°C, 15 psig (1 bar) for 20 minutes. Different models are available to accommodate either 25, 81 or 100 Cryovial tubes from 1.2 mL to 5 mL. A transparent cover allows the user to see the contents of the box, and is keyed to the base to prevent misalignment. Printed with a series of squares (numbered from 1 to 25, 1 to 42, 1 to 81, or 1 to 100), surface accepts writing with markers, facilitating inventory control. A unique color coding system uses colored plastic grids to separate the cover from the base on the 25, 42 and 81-place boxes. Those made to accept 100 tubes (series 2100) have a colored base instead of a grid. Removal of vials facilitated by an innovative vial picker supplied