Learn More
Andwin Scientific BOX DIVIDERS 9X9 12/PK
SDP
Catalog No. NC9079153
Encompass_Preferred
The Daigger Brand Cardboard Tube Storage Boxes have 81 cells that provide organization and division for 13 mm vials or 4.0 and 5.0 mL Cryovials. This product is sold in a pack of 12.
Catalog No. NC9079153 Supplier Andwin Scientific Supplier No. R79361MX2860HX
The Daigger Brand Cardboard Tube Storage Boxes have 81 cells that provide organization and division for 13 mm vials or 4.0 and 5.0 mL Cryovials. This product is sold in a pack of 12.