Learn More
Grainger WESTWARD Torque Wrench Head, 12-Point Box End, 10 mm, 9mm x 12mm Drive, 1 ea
Catalog No. 502793832
Encompass_Preferred
This 12-point box end interchangeable torque wrench head is designed for secure fastener engagement. The enclosed box end fits around the entire head of a fastener to provide a stable grip during torque application, minimizing slippage compared to open-end tools. Compatible with a range of interchangeable-head torque wrenches, this component helps consolidate equipment needs.
  • 12-point box end design for improved grip
  • Compatible with specified interchangeable-head torque wrenches
  • Constructed from alloy steel for durability
  • Satin finish
  • 10 mm head size
  • 99 ft-lb maximum torque
  • 0° head offset angle
Catalog No. 50-279-3832 Supplier Grainger Supplier No. 19ZC78
This 12-point box end interchangeable torque wrench head is designed for secure fastener engagement. The enclosed box end fits around the entire head of a fastener to provide a stable grip during torque application, minimizing slippage compared to open-end tools. Compatible with a range of interchangeable-head torque wrenches, this component helps consolidate equipment needs.
  • 12-point box end design for improved grip
  • Compatible with specified interchangeable-head torque wrenches
  • Constructed from alloy steel for durability
  • Satin finish
  • 10 mm head size
  • 99 ft-lb maximum torque
  • 0° head offset angle