Learn More
Grainger WESTWARD Torque Wrench Head, 15 mm, 9 x 12 mm Drive, 12-Point Box End, 1 ea
Catalog No. 502793810
Encompass_Preferred
This interchangeable torque wrench head is designed for precision fastening applications. Constructed from durable alloy steel, this tool features a 12-point box end configuration for secure engagement with fasteners.
  • Drive size of 9 mm x 12 mm
  • Size of 15 mm
  • Constructed from alloy steel
  • 12-point box end design
  • Satin finish
  • Overall length of 1 13/16 in.
  • Head offset angle of 0°
Catalog No. 50-279-3810 Supplier Grainger Supplier No. 19ZC83
This interchangeable torque wrench head is designed for precision fastening applications. Constructed from durable alloy steel, this tool features a 12-point box end configuration for secure engagement with fasteners.
  • Drive size of 9 mm x 12 mm
  • Size of 15 mm
  • Constructed from alloy steel
  • 12-point box end design
  • Satin finish
  • Overall length of 1 13/16 in.
  • Head offset angle of 0°