Learn More
Grainger Westward Torque Wrench Head, Alloy Steel, 9 mm x 12 mm Drive, 19 mm, 1 ea
Catalog No. 502793813
Encompass_Preferred
This interchangeable torque wrench head is designed for precision fastening applications. Constructed from durable alloy steel, this tool features a twelve-point box end configuration for reliable engagement with fasteners. It is engineered for compatibility with specific interchangeable torque wrenches, ensuring consistent performance in demanding mechanical tasks.
  • Constructed from high-quality alloy steel for enhanced durability
  • Features a 12-point box end design
  • Metric drive size of 9 mm x 12 mm
  • Satin finish for corrosion resistance
  • Overall length of 1 15/16 in.
  • Head size of 19 mm
Catalog No. 50-279-3813 Supplier Grainger Supplier No. 19ZC87
This interchangeable torque wrench head is designed for precision fastening applications. Constructed from durable alloy steel, this tool features a twelve-point box end configuration for reliable engagement with fasteners. It is engineered for compatibility with specific interchangeable torque wrenches, ensuring consistent performance in demanding mechanical tasks.
  • Constructed from high-quality alloy steel for enhanced durability
  • Features a 12-point box end design
  • Metric drive size of 9 mm x 12 mm
  • Satin finish for corrosion resistance
  • Overall length of 1 15/16 in.
  • Head size of 19 mm