Learn More
Grainger WESTWARD Torque Wrench Head, 12-Point Box End, 9mm x 12mm Drive, 22 mm, 1 ea
Catalog No. 502793896
Encompass_Preferred
This 12-point box end torque wrench head is designed for secure fastener engagement, fitting around the entire head of the fastener to provide a stable grip during operation. It is compatible with interchangeable-head torque wrenches, helping to reduce the necessity for multiple dedicated torque tools.
  • 12-point box end design
  • Constructed from durable alloy steel
  • Satin finish
  • 22 mm size
  • 9 mm x 12 mm drive size
  • 0° head offset angle
  • Overall length of 2 in.
Catalog No. 50-279-3896 Supplier Grainger Supplier No. 19ZC89
This 12-point box end torque wrench head is designed for secure fastener engagement, fitting around the entire head of the fastener to provide a stable grip during operation. It is compatible with interchangeable-head torque wrenches, helping to reduce the necessity for multiple dedicated torque tools.
  • 12-point box end design
  • Constructed from durable alloy steel
  • Satin finish
  • 22 mm size
  • 9 mm x 12 mm drive size
  • 0° head offset angle
  • Overall length of 2 in.