Learn More
Cole-Parmer Thermo Scientific Nalgene 5026-1010 PC Storage CryoBox, 100 x 1-2 mL Vials; 10/Pk
Catalog No. NC9537780 Shop All Cole-Parmer Products
Encompass
These nalgene cryogenic vial storage boxes are ideal for mechanical freezers or liquid nitrogen. They are made of polycarbonate and vented, allowing rapid cooling of contents. The boxes have a gridded insert and special lid with a one-way fit to make sample inventory foolproof. The boxes withstand temperatures from 196 to 121C and areautoclavable.
This item is not returnable. View return policy
Catalog No. NC9537780 Supplier Cole-Parmer Supplier No. 0675452
These nalgene cryogenic vial storage boxes are ideal for mechanical freezers or liquid nitrogen. They are made of polycarbonate and vented, allowing rapid cooling of contents. The boxes have a gridded insert and special lid with a one-way fit to make sample inventory foolproof. The boxes withstand temperatures from 196 to 121C and areautoclavable.