Learn More
World Precision Instrument Box Filament-14074
SDP
Catalog No. 502521508
Encompass
A glass puller is a laboratory instrument used to create fine, tapered glass micropipettes or microelectrodes. The choice of filament for the glass puller is crucial, as it directly influences the heating element's performance and, consequently, the quality of the pulled glass. Different types and shapes of filaments can be used, and the selection depends on factors such as the desired pipette. The box filament is configured in the shape of a square, allowing for controlled heating in the corners or along the edges of the filament. Similar to other specialized filament shapes, a box filament allows for controlled and localized heating during the glass softening and pulling process. Box filaments are often used when researchers require capillaries with specific shapes or features.
Catalog No. 50-252-1508 Supplier World Precision Instrument Supplier No. 14074
A glass puller is a laboratory instrument used to create fine, tapered glass micropipettes or microelectrodes. The choice of filament for the glass puller is crucial, as it directly influences the heating element's performance and, consequently, the quality of the pulled glass. Different types and shapes of filaments can be used, and the selection depends on factors such as the desired pipette. The box filament is configured in the shape of a square, allowing for controlled heating in the corners or along the edges of the filament. Similar to other specialized filament shapes, a box filament allows for controlled and localized heating during the glass softening and pulling process. Box filaments are often used when researchers require capillaries with specific shapes or features.